Assay Details
| Gene Symbol: LEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213788.1 | NP_998953.1 | 1 - 2 | 86 | 111 | |
| GenBank mRNA | AF516144.1 | - | 1 - 2 | 86 | 111 | |
| AJ409013.1 | - | 1 - 2 | 66 | 111 | ||
| AK233325.1 | - | 1953 | 111 | |||
More Information
Set Membership: |
|

LEAP2
Genomic Map