Assay Details
| Gene Symbol: DCPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213790.1 | NP_998955.1 | 1 - 2 | 203 | 87 | |
| XM_021102159.1 | XP_020957818.1 | 240 | 87 | |||
| GenBank mRNA | AK239804.1 | - | 1 - 2 | 220 | 87 | |
| AY040774.1 | - | 1 - 2 | 203 | 87 | ||
| AY610095.1 | - | 1 - 2 | 207 | 87 | ||
More Information
Set Membership: |
|

DCPS
Genomic Map