Assay Details
| Gene Symbol: MYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213791.2 | NP_998956.1 | 2 - 3 | 178 | 85 | |
| XM_005670649.3 | XP_005670706.1 | 348 | 85 | |||
| GenBank mRNA | AF513016.1 | - | 2 - 3 | 178 | 85 | |
| AJ487671.1 | - | 2 - 3 | 178 | 85 | ||
| AK344038.1 | - | 3 - 4 | 219 | 85 | ||
| JF829998.1 | - | 3 - 4 | 224 | 85 | ||
More Information
Set Membership: |
|

MYL2
Genomic Map