Assay Details
| Gene Symbol: CCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213833.2 | NP_998998.2 | 1 - 2 | 285 | 67 | |
| GenBank mRNA | U70060.1 | - | 1 - 2 | 274 | 67 | |
| U83916.1 | - | 1 - 2 | 274 | 67 | ||
More Information
Set Membership: |
|

CCN2
Genomic Map