Assay Details
| Gene Symbol: ATP2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213865.1 | NP_999030.1 | 17 - 18 | 2626 | 81 | |
| XM_021072075.1 | XP_020927734.1 | 3240 | 81 | |||
| GenBank mRNA | X15073.1 | - | 2626 | 81 | ||
| X15074.1 | - | 2626 | 81 | |||
More Information
Set Membership: |
|

ATP2A2
Genomic Map