Assay Details
| Gene Symbol: ATP2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213865.1 | NP_999030.1 | 19 - 20 | 2870 | 75 | |
| XM_021072075.1 | XP_020927734.1 | 3484 | 75 | |||
| GenBank mRNA | X15073.1 | - | 2870 | 75 | ||
| X15074.1 | - | 2870 | 75 | |||
More Information
Set Membership: |
|

ATP2A2
Genomic Map