Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213867.1 | NP_999032.1 | 1 - 2 | 150 | 101 | |
| GenBank mRNA | AK231005.1 | - | 1 - 2 | 150 | 101 | |
| AK391381.1 | - | 1 - 2 | 153 | 101 | ||
| AK391559.1 | - | 1 - 2 | 152 | 101 | ||
| M86923.1 | - | 1 - 2 | 150 | 101 | ||
| M99367.1 | - | 1 - 2 | 121 | 101 | ||
| X61151.1 | - | 141 | 101 | |||
More Information
Set Membership: |
|

CXCL8
Genomic Map