Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213867.1 | NP_999032.1 | 3 - 4 | 366 | 94 | |
| GenBank mRNA | AK231005.1 | - | 3 - 4 | 366 | 94 | |
| M86923.1 | - | 3 - 4 | 366 | 94 | ||
| M99367.1 | - | 3 - 4 | 337 | 94 | ||
More Information
Set Membership: |
|

CXCL8
Genomic Map