Assay Details
| Gene Symbol: SERPINE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213910.1 | NP_999075.1 | 8 | 1250 | 73 | |
| GenBank mRNA | AK234884.1 | - | 4 | 325 | 73 | |
| AK236245.1 | - | 8 | 1252 | 73 | ||
| Y11347.1 | - | 1250 | 73 | |||
More Information
Set Membership: |
|

SERPINE1
Genomic Map