Assay Details
| Gene Symbol: GALNS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213955.1 | NP_999120.1 | 11 | 1250 | 74 | |
| XM_021093500.1 | XP_020949159.1 | 1343 | 74 | |||
| XM_021093501.1 | XP_020949160.1 | 1192 | 74 | |||
| GenBank mRNA | AF322917.1 | - | 11 | 1250 | 74 | |
| AK237203.1 | - | 10 | 1384 | 74 | ||
More Information
Set Membership: |
|

GALNS
Genomic Map