Assay Details
| Gene Symbol: INSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213970.1 | NP_999135.1 | 2 | 349 | 139 | |
| GenBank mRNA | AK235810.1 | - | 2 | 356 | 139 | |
| AK348661.1 | - | 2 | 354 | 139 | ||
| AK351191.1 | - | 2 | 356 | 139 | ||
| X68369.1 | - | 349 | 139 | |||
More Information
Set Membership: |
|

INSL3
Genomic Map