Assay Details
| Gene Symbol: FTH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213975.1 | NP_999140.1 | 3 | 348 | 78 | |
| GenBank mRNA | D15071.1 | - | 3 | 348 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FTH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213975.1 | NP_999140.1 | 3 | 348 | 78 | |
| GenBank mRNA | D15071.1 | - | 3 | 348 | 78 | |
Set Membership: |
|