Assay Details
| Gene Symbol: SRD5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213988.1 | NP_999153.1 | 4 - 5 | 746 | 78 | |
| GenBank mRNA | AF008440.2 | - | 4 - 5 | 746 | 78 | |
More Information
Set Membership: |
|

SRD5A2
Genomic Map