Assay Details
| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214058.2 | NP_999223.1 | 2 - 3 | 662 | 65 | |
| XM_021079495.1 | XP_020935154.1 | 681 | 65 | |||
| GenBank mRNA | AF204929.1 | - | 2 - 3 | 663 | 65 | |
More Information
Set Membership: |
|

SERPINA7
Genomic Map