Assay Details
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214107.1 | NP_999272.1 | 2 - 3 | 240 | 79 | |
| GenBank mRNA | AK236575.1 | - | 2 - 3 | 262 | 79 | |
| AK395449.1 | - | 2 - 3 | 265 | 79 | ||
| D14875.1 | - | 2 - 3 | 240 | 79 | ||
More Information
Set Membership: |
|

ADM
Genomic Map