Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214118.2 | NP_999283.1 | 3 - 4 | 361 | 76 | |
| GenBank mRNA | AY116504.1 | - | 3 - 4 | 362 | 76 | |
| D21074.1 | - | 3 - 4 | 326 | 76 | ||
| DQ108393.1 | - | 3 - 4 | 326 | 76 | ||
| JX482567.1 | - | 1 - 2 | 77 | 76 | ||
| U61139.1 | - | 3 - 4 | 326 | 76 | ||
| U67175.1 | - | 3 - 4 | 326 | 76 | ||
More Information
Set Membership: |
|

CSF2
Genomic Map