Assay Details
| Gene Symbol: PRDX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214144.1 | NP_999309.1 | 2 - 3 | 323 | 58 | |
| GenBank mRNA | AF110735.2 | - | 2 - 3 | 323 | 58 | |
| AK231569.1 | - | 2 - 3 | 313 | 58 | ||
| AK233717.1 | - | 2 - 3 | 316 | 58 | ||
| AK234018.1 | - | 2 - 3 | 248 | 58 | ||
| AK392392.1 | - | 2 - 3 | 366 | 58 | ||
| AK399969.1 | - | 2 - 3 | 379 | 58 | ||
More Information
Set Membership: |
|

PRDX5
Genomic Map