Assay Details
| Gene Symbol: NPY2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214150.1 | NP_999315.1 | 1 | 1044 | 100 | |
| XM_005666792.3 | XP_005666849.1 | 2147 | 100 | |||
| GenBank mRNA | AF005780.2 | - | 1 | 1044 | 100 | |
| AK396399.1 | - | 2 | 1777 | 100 | ||
More Information
Set Membership: |
|

NPY2R
Genomic Map