Assay Details
| Gene Symbol: CCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214214.1 | NP_999379.1 | 1 - 2 | 129 | 58 | |
| GenBank mRNA | EU682382.1 | - | 1 - 2 | 77 | 58 | |
| X79416.1 | - | 129 | 58 | |||
More Information
Set Membership: |
|

CCL2
Genomic Map