Assay Details
| Gene Symbol: EDNRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214229.1 | NP_999394.1 | 3 - 4 | 799 | 71 | |
| XM_021100475.1 | XP_020956134.1 | 1274 | 71 | |||
| XM_021100476.1 | XP_020956135.1 | 942 | 71 | |||
| GenBank mRNA | S80652.1 | - | 3 - 4 | 799 | 71 | |
More Information
Set Membership: |
|

EDNRA
Genomic Map