Assay Details
| Gene Symbol: GNRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214273.1 | NP_999438.1 | 2 - 3 | 758 | 128 | |
| XM_005666725.3 | XP_005666782.1 | 128 | ||||
| GenBank mRNA | JN120792.1 | - | 2 - 3 | 128 | ||
| L29342.1 | - | 2 - 3 | 758 | 128 | ||
More Information
Set Membership: |
|

GNRHR
Genomic Map