Assay Details
| Gene Symbol: NPY1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214288.1 | NP_999453.1 | 1 - 2 | 701 | 149 | |
| XM_005666676.3 | XP_005666733.1 | 1426 | 149 | |||
| XM_005666677.3 | XP_005666734.1 | 1218 | 149 | |||
| GenBank mRNA | AF005779.2 | - | 1 - 2 | 701 | 149 | |
| AK238549.1 | - | 1 - 2 | 305 | 149 | ||
| AK395486.1 | - | 3 - 4 | 1155 | 149 | ||
More Information
Set Membership: |
|

NPY1R
Genomic Map