Assay Details
| Gene Symbol: APOE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214308.1 | NP_999473.1 | 3 - 4 | 280 | 63 | |
| GenBank mRNA | AK232315.1 | - | 3 - 4 | 300 | 63 | |
| AK399873.1 | - | 3 - 4 | 301 | 63 | ||
| X72835.1 | - | 280 | 63 | |||
More Information
Set Membership: |
|

APOE
Genomic Map