Assay Details
| Gene Symbol: SAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214358.1 | NP_999523.1 | 3 - 4 | 244 | 86 | |
| GenBank mRNA | AK240420.1 | - | 3 - 4 | 412 | 86 | |
| AK352180.1 | - | 3 - 4 | 381 | 86 | ||
| AK392196.1 | - | 3 - 4 | 432 | 86 | ||
| AY610311.1 | - | 3 - 4 | 363 | 86 | ||
| U57333.1 | - | 3 - 4 | 244 | 86 | ||
More Information
Set Membership: |
|

SAT1
Genomic Map