Assay Details
| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214380.2 | NP_999545.1 | 7 - 8 | 1341 | 66 | |
| GenBank mRNA | U90884.1 | - | 7 - 8 | 1342 | 66 | |
More Information
Set Membership: |
|

HMGCS2
Genomic Map