Assay Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214412.1 | NP_999577.1 | 2 - 3 | 961 | 77 | |
| XM_021098317.1 | XP_020953976.1 | 965 | 77 | |||
| GenBank mRNA | AB052254.1 | - | 2 - 3 | 1001 | 77 | |
| KP687263.1 | - | 2 - 3 | 960 | 77 | ||
| KU705620.1 | - | 1 - 2 | 840 | 77 | ||
More Information
Set Membership: |
|

CYP1A1
Genomic Map