Assay Details
| Gene Symbol: CYP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214432.1 | NP_999597.1 | 7 - 8 | 1102 | 74 | |
| XM_005667583.3 | XP_005667640.1 | 1078 | 74 | |||
| XM_005667584.3 | XP_005667641.1 | 997 | 74 | |||
| GenBank mRNA | AB009988.1 | - | 7 - 8 | 1102 | 74 | |
| AB529453.1 | - | 7 - 8 | 1075 | 74 | ||
| AF198112.1 | - | 5 - 6 | 796 | 74 | ||
| AK235908.1 | - | 7 - 8 | 1233 | 74 | ||
| AK394700.1 | - | 5 - 6 | 648 | 74 | ||
| AK397262.1 | - | 7 - 8 | 1257 | 74 | ||
| AY609713.1 | - | 7 - 8 | 1200 | 74 | ||
More Information
Set Membership: |
|

CYP51
Genomic Map