Assay Details
| Gene Symbol: SLA-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097431.1 | NP_001090900.1 | 8 | 1452 | 83 | |
| XM_021098437.1 | XP_020954096.1 | 1737 | 83 | |||
| GenBank mRNA | AK236791.1 | - | 9 | 1447 | 83 | |
| AK237112.1 | - | 10 | 1459 | 83 | ||
| AK390829.1 | - | 8 | 1451 | 83 | ||
| AK396028.1 | - | 9 | 1452 | 83 | ||
| AY135588.1 | - | 6 | 919 | 83 | ||
| AY135589.1 | - | 9 | 1406 | 83 | ||
| AY247766.1 | - | 8 | 1436 | 83 | ||
| AY459298.1 | - | 9 | 1417 | 83 | ||
| DQ303230.1 | - | 9 | 1421 | 83 | ||
| DQ992486.1 | - | 9 | 1449 | 83 | ||
| DQ992487.1 | - | 9 | 1450 | 83 | ||
| DQ992488.1 | - | 9 | 1449 | 83 | ||
| DQ992489.1 | - | 9 | 1448 | 83 | ||
| DQ992490.1 | - | 9 | 1449 | 83 | ||
| DQ992491.1 | - | 9 | 1449 | 83 | ||
| DQ992492.1 | - | 8 | 1452 | 83 | ||
| DQ992493.1 | - | 9 | 1401 | 83 | ||
| EF471311.1 | - | 8 | 1398 | 83 | ||
| EF472234.1 | - | 7 | 1171 | 83 | ||
| EU440342.1 | - | 8 | 1417 | 83 | ||
| EU440343.1 | - | 8 | 1417 | 83 | ||
| KU754553.1 | - | 8 | 1454 | 83 | ||
| Gene Symbol: LOC100513601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF464002.1 | - | 8 | 1488 | 83 | |
| AF464016.1 | - | 8 | 1488 | 83 | ||
| AK232064.1 | - | 8 | 1432 | 83 | ||
| AK232719.1 | - | 8 | 1435 | 83 | ||
| AK233366.1 | - | 8 | 1439 | 83 | ||
| AK237395.1 | - | 8 | 1435 | 83 | ||
| AK237474.1 | - | 8 | 1098 | 83 | ||
| AK237547.1 | - | 8 | 1201 | 83 | ||
| AK346139.1 | - | 8 | 1435 | 83 | ||
| AK346419.1 | - | 8 | 1435 | 83 | ||
| AK350415.1 | - | 8 | 1172 | 83 | ||
| AK351187.1 | - | 8 | 1424 | 83 | ||
| AK351887.1 | - | 8 | 1436 | 83 | ||
| AK391040.1 | - | 8 | 1460 | 83 | ||
| AK396599.1 | - | 8 | 1726 | 83 | ||
| AK396645.1 | - | 8 | 1215 | 83 | ||
| AK397821.1 | - | 8 | 1204 | 83 | ||
| AK398003.1 | - | 8 | 1440 | 83 | ||
| AK398067.1 | - | 8 | 1471 | 83 | ||
| AK398228.1 | - | 8 | 1436 | 83 | ||
| AK398945.1 | - | 8 | 1438 | 83 | ||
| AK400366.1 | - | 8 | 1438 | 83 | ||
| AY135591.1 | - | 8 | 1406 | 83 | ||
| AY135592.1 | - | 8 | 1405 | 83 | ||
| AY135593.1 | - | 8 | 1401 | 83 | ||
| AY135595.1 | - | 8 | 1406 | 83 | ||
| AY247764.1 | - | 8 | 1435 | 83 | ||
| AY247765.1 | - | 8 | 1436 | 83 | ||
| AY459297.1 | - | 8 | 1427 | 83 | ||
| AY459306.1 | - | 8 | 1427 | 83 | ||
| DQ303228.1 | - | 8 | 1406 | 83 | ||
| DQ303229.1 | - | 9 | 1406 | 83 | ||
| EF125045.1 | - | 8 | 1431 | 83 | ||
| EF125046.1 | - | 8 | 1431 | 83 | ||
| EF472233.1 | - | 8 | 1436 | 83 | ||
| EU170457.1 | - | 8 | 1406 | 83 | ||
| EU440334.1 | - | 8 | 1406 | 83 | ||
| EU440336.1 | - | 8 | 1406 | 83 | ||
| KU754545.1 | - | 8 | 1439 | 83 | ||
| KU754552.1 | - | 8 | 1439 | 83 | ||
| KU754557.1 | - | 8 | 1439 | 83 | ||
| Gene Symbol: LOC100622689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246245.1 | NP_001233174.1 | 8 | 1428 | 83 | |
| GenBank mRNA | AF464013.1 | - | 9 | 1337 | 83 | |
| AY247767.1 | - | 8 | 1437 | 83 | ||
| EF589959.1 | - | 8 | 1428 | 83 | ||
More Information
Set Membership: |
|

SLA-1
Genomic Map