Assay Details
| Gene Symbol: LOC100049687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098595.2 | NP_001092065.1 | 16 | 1749 | 132 | |
| GenBank mRNA | EF535028.1 | - | 16 | 1749 | 132 | |
More Information
Set Membership: |
|

LOC100049687
Genomic Map