Assay Details
| Gene Symbol: CHI3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001540.1 | NP_001001540.1 | 10 | 1651 | 126 | |
| GenBank mRNA | AK343786.1 | - | 10 | 1599 | 126 | |
| U19900.1 | - | 10 | 1651 | 126 | ||
| Z47803.1 | - | 1651 | 126 | |||
More Information
Set Membership: |
|

CHI3L1
Genomic Map