Assay Details
| Gene Symbol: IL23A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130236.1 | NP_001123708.1 | 2 - 3 | 277 | 116 | |
| GenBank mRNA | AB521204.1 | - | 2 - 3 | 420 | 116 | |
| KF246516.1 | - | 2 - 3 | 277 | 116 | ||
More Information
Set Membership: |
|

IL23A
Genomic Map