Assay Details
| Gene Symbol: ARPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134357.1 | NP_001127829.1 | 2 - 3 | 219 | 92 | |
| GenBank mRNA | AK390091.1 | - | 2 - 3 | 421 | 92 | |
| AY610438.1 | - | 2 - 3 | 300 | 92 | ||
| EU868838.1 | - | 2 - 3 | 219 | 92 | ||
More Information
Set Membership: |
|

ARPC5
Genomic Map