Assay Details
| Gene Symbol: TFPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135258.1 | NP_001128730.1 | 2 - 3 | 139 | 95 | |
| XM_005672009.3 | XP_005672066.1 | 436 | 95 | |||
| XM_013984326.2 | XP_013839780.1 | 582 | 95 | |||
| XM_013984328.2 | XP_013839782.1 | 651 | 95 | |||
| XM_013984329.2 | XP_013839783.1 | 508 | 95 | |||
| XM_021074527.1 | XP_020930186.1 | 437 | 95 | |||
| GenBank mRNA | EU090729.1 | - | 2 - 3 | 139 | 95 | |
More Information
Set Membership: |
|

TFPI
Genomic Map