Assay Details
| Gene Symbol: CXCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001623.1 | NP_001001623.1 | 2 | 1047 | 90 | |
| GenBank mRNA | AB119264.1 | - | 2 | 1047 | 90 | |
| AK344390.1 | - | 2 | 1030 | 90 | ||
More Information
Set Membership: |
|

CXCR6
Genomic Map