Assay Details
| Gene Symbol: MYH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104951.2 | NP_001098421.1 | 27 | 3950 | 71 | |
| XM_021066024.1 | XP_020921683.1 | 4394 | 71 | |||
| GenBank mRNA | AB025262.1 | - | 27 | 3953 | 71 | |
More Information
Set Membership: |
|

MYH1
Genomic Map