Assay Details
| Gene Symbol: LEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129967.1 | NP_001123439.1 | 12 | 2155 | 70 | |
| XM_005666937.3 | XP_005666994.1 | 3447 | 70 | |||
| XM_005666938.3 | XP_005666995.1 | 3141 | 70 | |||
| XM_005666939.3 | XP_005666996.1 | 3325 | 70 | |||
| XM_021100272.1 | XP_020955931.1 | 3441 | 70 | |||
| XM_021100273.1 | XP_020955932.1 | 3284 | 70 | |||
| XM_021100276.1 | XP_020955935.1 | 3159 | 70 | |||
| XM_021100277.1 | XP_020955936.1 | 2748 | 70 | |||
| XM_021100278.1 | XP_020955937.1 | 2441 | 70 | |||
| XM_021100279.1 | XP_020955938.1 | 2482 | 70 | |||
| XM_021100280.1 | XP_020955939.1 | 2412 | 70 | |||
| XM_021100281.1 | XP_020955940.1 | 2408 | 70 | |||
| GenBank mRNA | EU719607.1 | - | 12 | 2155 | 70 | |
More Information
Set Membership: |
|

LEF1
Genomic Map