Assay Details
| Gene Symbol: SRPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144113.1 | NP_001137585.1 | 16 | 2348 | 109 | |
| XM_005665912.3 | XP_005665969.1 | 2635 | 109 | |||
| XM_021098048.1 | XP_020953707.1 | 2007 | 109 | |||
| GenBank mRNA | AK240278.1 | - | 6 | 628 | 109 | |
| FJ476290.1 | - | 16 | 2348 | 109 | ||
| FJ610142.1 | - | 16 | 2040 | 109 | ||
More Information
Set Membership: |
|

SRPK1
Genomic Map