Assay Details
| Gene Symbol: POPDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144114.1 | NP_001137586.1 | 4 | 1952 | 111 | |
| XM_021068245.1 | XP_020923904.1 | 1922 | 111 | |||
| GenBank mRNA | EU334862.1 | - | 4 | 1952 | 111 | |
| EU334863.1 | - | 4 | 1811 | 111 | ||
| EU334864.1 | - | 3 | 927 | 111 | ||
More Information
Set Membership: |
|

POPDC2
Genomic Map