Assay Details
| Gene Symbol: CD82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145218.2 | NP_001138690.1 | 7 | 734 | 102 | |
| XM_021077185.1 | XP_020932844.1 | 594 | 102 | |||
| GenBank mRNA | AK232533.1 | - | 7 | 696 | 102 | |
| AK236958.1 | - | 131 | 102 | |||
| AM498376.1 | - | 6 | 555 | 102 | ||
| CK451293.1 | - | 2 | 219 | 102 | ||
More Information
Set Membership: |
|

CD82
Genomic Map