Assay Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145221.1 | NP_001138693.1 | 6 | 553 | 69 | |
| GenBank mRNA | AK233864.1 | - | 6 | 568 | 69 | |
| AK399000.1 | - | 6 | 572 | 69 | ||
| FJ455237.1 | - | 6 | 553 | 69 | ||
More Information
Set Membership: |
|

ARF5
Genomic Map