Assay Details
| Gene Symbol: ACTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164650.1 | NP_001158122.1 | 1 - 2 | 165 | 84 | |
| XM_013983522.2 | XP_013838976.1 | 245 | 84 | |||
| XM_021072186.1 | XP_020927845.1 | 152 | 84 | |||
| GenBank mRNA | AK234691.1 | - | 2 - 3 | 191 | 84 | |
| AK240348.1 | - | 2 - 3 | 395 | 84 | ||
| AK394915.1 | - | 2 - 3 | 439 | 84 | ||
| AK394958.1 | - | 2 - 3 | 342 | 84 | ||
| AK398905.1 | - | 3 - 4 | 245 | 84 | ||
| FJ547477.1 | - | 1 - 2 | 165 | 84 | ||
More Information
Set Membership: |
|

ACTA2
Genomic Map