Assay Details
| Gene Symbol: PHKG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166317.2 | NP_001159789.2 | 6 - 7 | 659 | 78 | |
| XM_005652997.3 | XP_005653054.1 | 710 | 78 | |||
| XM_013992337.2 | XP_013847791.1 | 478 | 78 | |||
| XM_021085628.1 | XP_020941287.1 | 480 | 78 | |||
| GenBank mRNA | EU169240.1 | - | 6 - 7 | 615 | 78 | |
| KJ186785.1 | - | 5 - 6 | 556 | 78 | ||
More Information
Set Membership: |
|

PHKG2
Genomic Map