Assay Details
| Gene Symbol: CCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166491.1 | NP_001159963.1 | 1 - 2 | 127 | 94 | |
| GenBank mRNA | GQ856244.1 | - | 1 - 2 | 127 | 94 | |
More Information
Set Membership: |
|

CCL1
Genomic Map