Assay Details
| Gene Symbol: ELOVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167634.1 | NP_001161106.1 | 2 - 3 | 233 | 67 | |
| XM_005671398.2 | XP_005671455.1 | 328 | 67 | |||
| GenBank mRNA | AB529460.1 | - | 2 - 3 | 233 | 67 | |
| AK238786.1 | - | 2 - 3 | 328 | 67 | ||
| AK351721.1 | - | 2 - 3 | 328 | 67 | ||
More Information
Set Membership: |
|

ELOVL3
Genomic Map