Assay Details
| Gene Symbol: RHOJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168417.1 | NP_001161889.1 | 4 - 5 | 493 | 63 | |
| GenBank mRNA | AB530147.1 | - | 4 - 5 | 493 | 63 | |
| AK235956.1 | - | 3 - 4 | 839 | 63 | ||
| AK236185.1 | - | 4 - 5 | 898 | 63 | ||
More Information
Set Membership: |
|

RHOJ
Genomic Map