Assay Details
| Gene Symbol: FSCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146300.2 | NP_001139772.1 | 1 - 2 | 951 | 63 | |
| GenBank mRNA | AK236699.1 | - | 1 - 2 | 953 | 63 | |
| AK392031.1 | - | 1 - 2 | 949 | 63 | ||
| FJ790418.1 | - | 1 - 2 | 953 | 63 | ||
More Information
Set Membership: |
|

FSCN1
Genomic Map