Assay Details
| Gene Symbol: SMYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160089.1 | NP_001153561.1 | 9 - 10 | 1349 | 54 | |
| XM_021085617.1 | XP_020941276.1 | 1366 | 54 | |||
| GenBank mRNA | EU661947.1 | - | 9 - 10 | 1349 | 54 | |
More Information
Set Membership: |
|

SMYD1
Genomic Map