Assay Details
| Gene Symbol: SPATA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177908.1 | NP_001171379.1 | 4 - 5 | 373 | 82 | |
| XM_013978347.2 | XP_013833801.1 | 420 | 82 | |||
| XM_013978348.2 | XP_013833802.1 | 325 | 82 | |||
| XM_013978349.2 | XP_013833803.1 | 237 | 82 | |||
| XM_021098001.1 | XP_020953660.1 | 420 | 82 | |||
| GenBank mRNA | AK347650.1 | - | 4 - 5 | 373 | 82 | |
| AK393266.1 | - | 4 - 5 | 400 | 82 | ||
| GU475000.1 | - | 4 - 5 | 373 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013978349.2 transcripts.
Set Membership: |
|

SPATA7
Genomic Map