Assay Details
| Gene Symbol: CPEB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185049.2 | NP_001171978.2 | 8 - 9 | 2787 | 85 | |
| XM_021100320.1 | XP_020955979.1 | 2811 | 85 | |||
| XM_021100322.1 | XP_020955981.1 | 2730 | 85 | |||
| XM_021100323.1 | XP_020955982.1 | 2721 | 85 | |||
| XM_021100324.1 | XP_020955983.1 | 2706 | 85 | |||
| XM_021100325.1 | XP_020955984.1 | 2697 | 85 | |||
| XM_021100326.1 | XP_020955985.1 | 1229 | 85 | |||
| XM_021100327.1 | XP_020955986.1 | 4246 | 85 | |||
| GenBank mRNA | HM037344.1 | - | 7 - 8 | 852 | 85 | |
| HM037345.1 | - | 7 - 8 | 843 | 85 | ||
More Information
Set Membership: |
|

CPEB2
Genomic Map