Assay Details
| Gene Symbol: ARPC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185134.1 | NP_001172063.1 | 3 - 4 | 405 | 81 | |
| GenBank mRNA | AK235432.1 | - | 3 - 4 | 495 | 81 | |
| AY609812.1 | - | 3 - 4 | 504 | 81 | ||
| EU868839.1 | - | 3 - 4 | 405 | 81 | ||
More Information
Set Membership: |
|

ARPC5L
Genomic Map